7-Fluorobenzo[b]thiophene 95%

Supplier: Ambeed
Warning

A129203-1G A129203-250MG
76983-586EA 181.42 USD
76983-586 76983-588
7-Fluorobenzo[b]thiophene 95%
7-Fluorobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅FS
MW: 152.189 g/mol
MDL Number: MFCD17169311
CAS Number: 346592-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR