6-Bromo-4-methyl-1,2,3,4-tetrahydroquinoline 97%
Supplier: Ambeed
|
Warning
|
A129173-100MG
A129173-10G
A129173-1G
A129173-250MG
A129173-5G
77009-826EA
27.59
USD
77009-826
77009-828
77009-830
77009-832
77009-834
6-Bromo-4-methyl-1,2,3,4-tetrahydroquinoline 97%
6-Bromo-4-methyl-1,2,3,4-tetrahydroquinoline
Precaución: for Research Use Only
Formula:
C₁₀H₁₂BrN MW: 226.11 g/mol |
MDL Number:
MFCD26626379 CAS Number: 946837-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...