6-Chloroimidazo[1,2-b]pyridazin-8-amine 95%

Supplier: Ambeed
Warning

A129079-100MG A129079-1G A129079-250MG A129079-5G
76983-578EA 229.16 USD
76983-578 76983-580 76983-582 76983-584
6-Chloroimidazo[1,2-b]pyridazin-8-amine 95%
6-Chloroimidazo[1,2-b]pyridazin-8-amine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
MW: 168.58 g/mol
MDL Number: MFCD21336327
CAS Number: 1161847-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR