6-Chloro-N4-methylpyrimidine-2,4-diamine 97%

Supplier: Ambeed
Warning

A129036-1G A129036-250MG A129036-5G
77009-820EA 51 USD
77009-820 77009-822 77009-824
6-Chloro-N4-methylpyrimidine-2,4-diamine 97%
6-Chloro-N4-methylpyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₅H₇ClN₄
MDL Number: MFCD02091108
CAS Number: 1005-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR