4-(4-Chlorothiazol-2-yl)morpholine 95%
Supplier: Ambeed
|
Warning
|
A128990-10G
A128990-1G
A128990-5G
77026-966EA
1287.94
USD
77026-966
77026-968
77026-970
4-(4-Chlorothiazol-2-yl)morpholine 95%
4-(4-Chlorothiazol-2-yl)morpholine
Precaución: for Research Use Only
Formula:
C₇H₉ClN₂OS MW: 204.68 g/mol |
MDL Number:
MFCD09909639 CAS Number: 848841-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...