2-Chloro-5-iodo-4-pyridinamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-chloro-5-iodopyridine

A128869-10G A128869-1G A128869-250MG A128869-25G A128869-5G
77026-956EA 138.98 USD
77026-956 77026-958 77026-960 77026-962 77026-964
2-Chloro-5-iodo-4-pyridinamine 95%
2-Chloro-5-iodo-4-pyridinamine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MW: 254.455 g/mol
MDL Number: MFCD09834661
CAS Number: 800402-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR