N-Fmoc-N'-trityl-D-asparagine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Asn(Trt)-OH

A128604-100G A128604-10G A128604-1G A128604-250MG A128604-25G A128604-5G
76996-224EA 292 USD
76996-224 76996-226 76996-228 76996-230 76996-232 76996-234
N-Fmoc-N'-trityl-D-asparagine 97%
N-Fmoc-N'-trityl-D-asparagine

Precaución: for Research Use Only

Formula: C₃₈H₃₂N₂O₅
MDL Number: MFCD00151919
CAS Number: 180570-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR