5-Bromoindole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1H-indole

A128553-100G A128553-10G A128553-25G A128553-500G
77009-810EA 20.16 USD
77009-810 77009-812 77009-814 77009-816
5-Bromoindole 97%
5-Bromoindole

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD00005670
CAS Number: 10075-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR