5-Bromo-2-methoxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

A128539-100G A128539-10G A128539-1G A128539-25G A128539-5G
77009-800EA 161.26 USD
77009-800 77009-802 77009-804 77009-806 77009-808
5-Bromo-2-methoxy-3-nitropyridine 97%
5-Bromo-2-methoxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₃
MW: 233.0200 g/mol
MDL Number: MFCD07374968
CAS Number: 152684-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR