5-Fluoro-3,4-dihydro-2,4-dioxo-1(2H)-Pyrimidineacetic Acid 97%

Supplier: Ambeed
Warning

A128430-1G A128430-250MG A128430-5G
77009-792EA 152.01 USD
77009-792 77009-794 77009-796
5-Fluoro-3,4-dihydro-2,4-dioxo-1(2H)-Pyrimidineacetic Acid 97%
5-Fluoro-3,4-dihydro-2,4-dioxo-1(2H)-Pyrimidineacetic Acid

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₄
MDL Number: MFCD00832183
CAS Number: 56059-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR