2-Bromo-3-chloro-5-fluoropyridine 95%

Supplier: Ambeed
Warning

A128376-1G A128376-250MG A128376-5G A128376-25G
76983-540EA 13 USD
76983-540 76983-542 76983-544 77885-131
2-Bromo-3-chloro-5-fluoropyridine 95%
2-Bromo-3-chloro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MDL Number: MFCD13185350
CAS Number: 1214326-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR