Ethyl 4-(5-amino-1-methyl-1H-1,3-benzimidazol-2-yl)butanoate 98%

Supplier: Ambeed
Warning

Synonyms: Bendamustine Impurity 19, Ethyl 4-(5-amino-1-methyl-1H-benzo[d]imidazol-2-yl)butanoate

A128291-250MG A128291-1G A128291-100G
77026-934EA 8 USD
77026-934 77666-834 77871-588
Ethyl 4-(5-amino-1-methyl-1H-1,3-benzimidazol-2-yl)butanoate 98%
Ethyl 4-(5-amino-1-methyl-1H-1,3-benzimidazol-2-yl)butanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₉N₃O₂
MDL Number: MFCD09840909
CAS Number: 3543-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR