5-Chloro-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A128251-1G A128251-250MG A128251-5G
77009-786EA 70.02 USD
77009-786 77009-788 77009-790
5-Chloro-1H-indazole-3-carboxylic acid 97%
5-Chloro-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MW: 196.59 g/mol
MDL Number: MFCD02326712
CAS Number: 1077-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR