5,7-Dichloroquinoline 98%
Supplier: Ambeed
|
Warning
|
A128204-100MG
A128204-1G
A128204-250MG
A128204-5G
77026-926EA
14.86
USD
77026-926
77026-928
77026-930
77026-932
5,7-Dichloroquinoline 98%
5,7-Dichloroquinoline
Precaución: for Research Use Only
Formula:
C₉H₅Cl₂N MW: 198.0500 g/mol |
MDL Number:
MFCD11504933 CAS Number: 4964-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...