4-Bromo-6-methylpicolinic acid 95%

Supplier: Ambeed
Warning

A128188-100MG A128188-250MG
76983-534EA 99.73 USD
76983-534 76983-536
4-Bromo-6-methylpicolinic acid 95%
4-Bromo-6-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD07374944
CAS Number: 886372-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR