4-Bromo-2-chloropyridin-3-amine 95%

Supplier: Ambeed
Warning

A128156-100MG A128156-1G A128156-250MG
76983-532EA 31 USD
76983-532 77611-118 77889-885
4-Bromo-2-chloropyridin-3-amine 95%
4-Bromo-2-chloropyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MDL Number: MFCD21025849
CAS Number: 1354021-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR