2-(Dimethylamino)-2-oxoethyl 2-(4-hydroxyphenyl)acetate 98%

Supplier: Ambeed
Warning

A127936-100G A127936-1G A127936-25G A127936-5G
77009-774EA 248.26 USD
77009-774 77009-776 77009-778 77009-780
2-(Dimethylamino)-2-oxoethyl 2-(4-hydroxyphenyl)acetate 98%
2-(Dimethylamino)-2-oxoethyl 2-(4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MW: 237.25 g/mol
MDL Number: MFCD09952449
CAS Number: 59721-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR