(2,3-Dimethylphenyl)(1H-imidazol-4-yl)methanone 98%

Supplier: Ambeed
Warning

A127855-100MG A127855-5G A127855-1G
76983-528EA 90.18 USD
76983-528 77611-110 77683-134
(2,3-Dimethylphenyl)(1H-imidazol-4-yl)methanone 98%
(2,3-Dimethylphenyl)(1H-imidazol-4-yl)methanone

Precaución: For research use only.

Formula: C₁₂H₁₂N₂O
MW: 200.23 g/mol
MDL Number: MFCD21332884
CAS Number: 91874-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR