10,11-Dihydrocarbamazepine 98%

Supplier: Ambeed
Warning

Synonyms: 10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carboxamide

A127850-100MG A127850-1G A127850-250MG A127850-5G
77009-766EA 75.33 USD
77009-766 77009-768 77009-770 77683-133
10,11-Dihydrocarbamazepine 98%
10,11-Dihydrocarbamazepine

Precaución: For research use only.

Formula: C₁₅H₁₄N₂O
MW: 238.29 g/mol
MDL Number: MFCD00005072
CAS Number: 3564-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR