Methyl-2-methoxynicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methoxynicotinate

A127786-100G A127786-10G A127786-25G A127786-5G
76996-178EA 98 USD
76996-178 76996-180 76996-182 76996-184
Methyl-2-methoxynicotinate 97%
Methyl-2-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD06801052
CAS Number: 67367-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR