Methyl 2,4,5-trifluorobenzoate 98%

Supplier: Ambeed
Warning

A127707-100G A127707-1G A127707-250MG A127707-25G A127707-5G
77009-750EA 290.69 USD
77009-750 77009-752 77009-754 77009-756 77009-758
Methyl 2,4,5-trifluorobenzoate 98%
Methyl 2,4,5-trifluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MW: 190.119 g/mol
MDL Number: MFCD06203702
CAS Number: 20372-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR