2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A127699-10G
A127699-1G
A127699-250MG
A127699-5G
77026-910EA
338.43
USD
77026-910
77026-912
77026-914
77026-916
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine 97%
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₃ClIN₃ MW: 279.4700 g/mol |
MDL Number:
MFCD13189363 CAS Number: 1060815-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...