trans-4-Aminoadamantan-1-ol 95%

Supplier: Ambeed
Warning

A127634-10G A127634-1G A127634-5G
77026-898EA 290.69 USD
77026-898 77026-900 77026-902
trans-4-Aminoadamantan-1-ol 95%
trans-4-Aminoadamantan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO
MW: 167.248 g/mol
MDL Number: MFCD12761603
CAS Number: 62058-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR