1-Boc-4-(4-bromophenoxy)piperidine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(4-Bromophenoxy)piperidine

A127548-100MG A127548-1G A127548-250MG A127548-5G
77026-886EA 16 USD
77026-886 77026-888 77026-890 77026-892
1-Boc-4-(4-bromophenoxy)piperidine 95%
1-Boc-4-(4-bromophenoxy)piperidine

Precaución: for Research Use Only

Formula: C₁₆H₂₂BrNO₃
MW: 356.2548 g/mol
MDL Number: MFCD11053889
CAS Number: 308386-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR