4-Amino-3-fluoropyridine 98%

Supplier: Ambeed
Warning

A127484-1G A127484-250MG A127484-10G
76996-162EA 19 USD
76996-162 77670-808 77881-015
4-Amino-3-fluoropyridine 98%
4-Amino-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
MDL Number: MFCD00234072
CAS Number: 2247-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR