7-Bromo-5-nitroindoline 97%

Supplier: Ambeed
Warning

A127423-1G A127423-5G
76983-508EA 25 USD
76983-508 77889-881
7-Bromo-5-nitroindoline 97%
7-Bromo-5-nitroindoline

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O₂
MDL Number: MFCD09258800
CAS Number: 87240-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR