Methyl 3-chloro-5-cyanobenzoate 95%

Supplier: Ambeed
Warning

A127310-100MG A127310-1G A127310-25G
77009-732EA 9 USD
77009-732 77009-734 77009-736
Methyl 3-chloro-5-cyanobenzoate 95%
Methyl 3-chloro-5-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MDL Number: MFCD11976045
CAS Number: 327056-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR