3-Bromo-2-methoxybenzamide 95%

Supplier: Ambeed
Warning

A127195-100MG A127195-1G A127195-250MG A127195-5G
77009-714EA 14.86 USD
77009-714 77009-716 77009-718 77009-720
3-Bromo-2-methoxybenzamide 95%
3-Bromo-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD18391870
CAS Number: 791136-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR