3-Boc-amino-4-bromopyridine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Boc-Amino-4-bromopyridine

A126969-100MG A126969-250MG
76983-492EA 36.08 USD
76983-492 76983-494
3-Boc-amino-4-bromopyridine 95%
3-Boc-amino-4-bromopyridine

Precaución: for Research Use Only

Formula: C10H13BrN2O2
MW: 273.1300 g/mol
MDL Number: MFCD05664029
CAS Number: 885275-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR