4,6-Dimethyl-3-nitropyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A126963-1G A126963-250MG A126963-5G
76996-156EA 67 USD
76996-156 76996-158 76996-160
4,6-Dimethyl-3-nitropyridin-2(1H)-one 97%
4,6-Dimethyl-3-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD06809625
CAS Number: 22934-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR