2-Bromo-3-methyl-4-(trifluoromethyl)pyridine 95%
Supplier: Ambeed
|
Danger
|
A126891-1G
A126891-100MG
A126891-250MG
77009-712EA
189
USD
77009-712
77889-877
77889-878
2-Bromo-3-methyl-4-(trifluoromethyl)pyridine 95%
2-Bromo-3-methyl-4-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₇H₅BrF₃N |
MDL Number:
MFCD26516444 CAS Number: 1448776-78-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...