Enalapril maleate 98%

Supplier: Ambeed
Warning

Synonyms: 1-[N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanyl]-L-proline Maleate, (S)-1-((S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoyl)pyrrolidine-2-carboxylic acid Maleate

A126648-10G A126648-25G
77026-868EA 8.49 USD
77026-868 77026-870
Enalapril maleate 98%
Enalapril maleate

Precaución: for Research Use Only

Formula: C₂₀H₂₈N₂O₅
MW: 492.53 g/mol
MDL Number: MFCD00133304
CAS Number: 76095-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR