1-Methylcyclopropane-1-sulfonamide 97%

Supplier: Ambeed
Warning

A126645-100MG A126645-10G A126645-1G A126645-250MG A126645-25G A126645-5G
77026-856EA 9.55 USD
77026-856 77026-858 77026-860 77026-862 77026-864 77026-866
1-Methylcyclopropane-1-sulfonamide 97%
1-Methylcyclopropane-1-sulfonamide

Precaución: for Research Use Only

Formula: C₄H₉NO₂S
MW: 135.1900 g/mol
MDL Number: MFCD11501466
CAS Number: 669008-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR