4-Bromothiazole-2-carbonitrile 97%

Supplier: Ambeed
Warning

A126638-100MG A126638-5G A126638-250MG A126638-1G
77009-682EA 47.75 USD
77009-682 77009-684 77592-454 77611-090
4-Bromothiazole-2-carbonitrile 97%
4-Bromothiazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₄HBrN₂S
MW: 189.03 g/mol
MDL Number: MFCD09607690
CAS Number: 1017781-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR