Piperine 97%

Supplier: Ambeed
Warning

Synonyms: (2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one, 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine

A126615-100G A126615-10G A126615-1G A126615-1MG A126615-250MG A126615-25G A126615-5G
77026-842EA 332.07 USD
77026-842 77026-844 77026-846 77026-848 77026-850 77026-852 77026-854
Piperine 97%
Piperine

Precaución: for Research Use Only

Formula: C₁₇H₁₉NO₃
MW: 285.34 g/mol
MDL Number: MFCD00005839
CAS Number: 94-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR