2-Amino-4-morpholin-4-ylpyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-morpholin-4-yl-pyrimidine

A126597-10G A126597-1G A126597-250MG
77026-836EA 321.46 USD
77026-836 77026-838 77026-840
2-Amino-4-morpholin-4-ylpyrimidine 98%
2-Amino-4-morpholin-4-ylpyrimidine

Precaución: for Research Use Only

Formula: C₈H₁₂N₄O
MW: 180.21 g/mol
MDL Number: MFCD11520888
CAS Number: 861031-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR