Ethyl 2,4-diaminopyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A126585-1G A126585-250MG
77009-678EA 31 USD
77009-678 77009-680
Ethyl 2,4-diaminopyrimidine-5-carboxylate 97%
Ethyl 2,4-diaminopyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₄O₂
MDL Number: MFCD04973411
CAS Number: 15400-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR