3-((2S,4S)-4-Mercapto-1-(((4-nitrobenzyl)oxy)carbonyl)pyrrolidine-2-carboxamido)benzoic acid 97%

Supplier: Ambeed
Warning

A126495-100MG A126495-1G A126495-1MG A126495-250MG
76996-142EA 18.04 USD
76996-142 76996-144 76996-146 76996-148
3-((2S,4S)-4-Mercapto-1-(((4-nitrobenzyl)oxy)carbonyl)pyrrolidine-2-carboxamido)benzoic acid 97%
3-((2S,4S)-4-Mercapto-1-(((4-nitrobenzyl)oxy)carbonyl)pyrrolidine-2-carboxamido)benzoic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₃O₇S
MW: 445.45 g/mol
MDL Number: MFCD09263754
CAS Number: 202467-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR