Tri-o-tolyl phosphite 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A126449-1G A126449-250MG A126449-100G A126449-5G
76983-482EA 33 USD
76983-482 77683-123 77885-122 77885-123
Tri-o-tolyl phosphite 95%, Ambeed.Inc
Tri-o-tolyl phosphite

Precaución: For research use only.

Formula: C₂₁H₂₁O₃P
MDL Number: MFCD00014911
CAS Number: 2622-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR