1-(5-Methyl-2-nitrophenyl)ethanone 97%

Supplier: Ambeed
Warning

A126446-10G A126446-1G
76983-478EA 388.29 USD
76983-478 76983-480
1-(5-Methyl-2-nitrophenyl)ethanone 97%
1-(5-Methyl-2-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.17 g/mol
MDL Number: MFCD11046202
CAS Number: 69976-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR