Thieno[2,3-b]pyridin-5-amine 98%

Supplier: Ambeed
Warning

A126333-100MG A126333-250MG
77026-832EA 99.73 USD
77026-832 77026-834
Thieno[2,3-b]pyridin-5-amine 98%
Thieno[2,3-b]pyridin-5-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD14706798
CAS Number: 21344-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR