1,3-Dichloro-2-fluoro-5-iodobenzene 97%

Supplier: Ambeed
Warning

A126253-1G A126253-250MG
76983-474EA 43.5 USD
76983-474 76983-476
1,3-Dichloro-2-fluoro-5-iodobenzene 97%
1,3-Dichloro-2-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FI
MW: 290.89 g/mol
MDL Number: MFCD09996900
CAS Number: 133307-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR