tert-Butyl 4-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 96%

Supplier: Ambeed
Warning

A126162-1G A126162-250MG A126162-5G
76983-468EA 180.36 USD
76983-468 76983-470 76983-472
tert-Butyl 4-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 96%
tert-Butyl 4-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MW: 297.15 g/mol
MDL Number: MFCD20527472
CAS Number: 1228014-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR