N-(Diphenylmethylene)glycine ethyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl (diphenylmethylenamino)acetate, Ethyl 2-((diphenylmethylene)amino)acetate, Ethyl-2-((diphenylmethylene)amino)acetate, Ethyl N-(diphenylmethylene)glycinate
A126045-1000G
A126045-100G
A126045-10G
A126045-25G
A126045-500G
A126045-5G
77026-804EA
241.89
USD
77026-804
77026-806
77026-808
77026-810
77026-812
77026-814
N-(Diphenylmethylene)glycine ethyl ester 95%
N-(Diphenylmethylene)glycine ethyl ester
Precaución: for Research Use Only
Formula:
C₁₇H₁₇NO₂ MW: 267.32 g/mol |
MDL Number:
MFCD00010590 CAS Number: 69555-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...