(S)-3-Methyl-1-(2-piperidine-1-yl-phenyl)-butylamine 97%

Supplier: Ambeed
Warning

A126026-10G A126026-1G A126026-250MG A126026-5G A126026-25G
76996-120EA 498 USD
76996-120 76996-122 76996-124 77670-786 77881-010
(S)-3-Methyl-1-(2-piperidine-1-yl-phenyl)-butylamine 97%
(S)-3-Methyl-1-(2-piperidine-1-yl-phenyl)-butylamine

Precaución: for Research Use Only

Formula: C₁₆H₂₆N₂
MDL Number: MFCD03839861
CAS Number: 147769-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR