4-Amino-2-bromo-6-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methylpyridin-4-amine, 4-Amino-6-bromo-2-picoline

A126007-100G A126007-100MG A126007-10G A126007-1G A126007-250MG A126007-25G A126007-5G
77026-778EA 2789.11 USD
77026-778 77026-780 77026-782 77026-784 77026-786 77026-788 77026-790
4-Amino-2-bromo-6-methylpyridine 97%
4-Amino-2-bromo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
MDL Number: MFCD12400760
CAS Number: 79055-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR