3-(Methylsulfonyl)-1-propanol 97%

Supplier: Ambeed
Warning

A125986-100G A125986-10G A125986-1G A125986-25G A125986-5G
76996-110EA 135 USD
76996-110 76996-112 76996-114 76996-116 76996-118
3-(Methylsulfonyl)-1-propanol 97%
3-(Methylsulfonyl)-1-propanol

Precaución: for Research Use Only

Formula: C₄H₁₀O₃S
MDL Number: MFCD01696727
CAS Number: 2058-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR