5-Bromo-4-chloro-6-methylpyrimidin-2-amine 98%

Supplier: Ambeed
Warning

A125955-1G A125955-5G
77026-776EA 22 USD
77026-776 77599-824
5-Bromo-4-chloro-6-methylpyrimidin-2-amine 98%
5-Bromo-4-chloro-6-methylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₅H₅BrClN₃
MDL Number: MFCD00275421
CAS Number: 6314-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR