tert-Butyl 3-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate 97%

Supplier: Ambeed
Warning

A125733-100MG A125733-250MG
77009-628EA 18.04 USD
77009-628 77009-630
tert-Butyl 3-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate 97%
tert-Butyl 3-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MW: 297.15 g/mol
MDL Number: MFCD16877631
CAS Number: 192189-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR