3,4,5-Trifluorophenol 98%

Supplier: Ambeed
Danger

A125691-25G A125691-5G
77026-774EA 21.22 USD
77026-774 77571-534
3,4,5-Trifluorophenol 98%
3,4,5-Trifluorophenol

Precaución: for Research Use Only

Formula: C₆H₃F₃O
MW: 148.08 g/mol
MDL Number: MFCD00042427
CAS Number: 99627-05-1
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR